Learn More
Medchemexpress LLC Iohexol | 66108-95-0 | 99.5% | 1 ML
SDP
Supplier:  Medchemexpress LLC HYB059410MM/1ML
Encompass_Preferred
Iohexol is a non-ionic, water-soluble contrast agent that serves as a reference marker for determining glomerular filtration rate (GFR) at the level of renal function.
  • Can be used for contrast in myelography.
  • Suitable for computerized tomography (cisternography, ventriculography).
  • Applicable for MicroCT imaging.
Catalog No. 50-004-08361