Learn More
Medchemexpress LLC Iomeprol intermediat 10g
SDP
Supplier:  Medchemexpress LLC HYW09701810G
Encompass_Preferred
Iomeprol intermediate-1 (compound 5) is an iodinated polysaccharide compound used in the synthesis of contrast agents The amino group of Iomeprol intermediate-1 can be coupled to the carboxyl group of the D-glucuronide subunit of non-animal stable hyaluronic acid (NASHA) providing a water-soluble radiopaque moiety[1]
Catalog No. 50-002-05159