Learn More
Abcam ION Potassium Green-2 TMA+ Salt, K+ indicator, 500UG
Supplier:  Abcam AB142807500UG
Encompass_Preferred
MW 1131.24 g/mol, Purity >90%. Cell membrane impermeable K+ indicator. Water soluble. Related product (ab142806). Fluorescent probe. Active in vitro.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-258-3310