Learn More
Thermo Scientific Ion Transfer Tube High 1 EA
Supplier:  Thermo Scientific 8050020045
Encompass
Stainless steel ion transfer tube with 1/4 turn fitting, 2.5mm diameter, 0.6 x 1.6mm dimensions for ion transmission in analytical instrumentation to enable ion analysis
Catalog No. NC1502681