Learn More
BIOGEMS INTERNATIONAL INC Ionomycin Calcium Salt 10mg
SDP
Supplier:  BIOGEMS INTERNATIONAL INC 560821210MG
Encompass
Ionomycin is an ionphore discovered in Streptomyces conglobatus and used in the study of intracellular calcium. It is also reported to stimulate the intracellular production of IL-2, IL-4 and perforin. It is supplied as a a calcium salt for ease of storage.
Catalog No. 50-153-0187