Learn More
Cayman Chemical Ionomycin (calcium salt), 56092-82-1, 10 mg
SDP
Supplier:  Cayman Chemical 1193210
Encompass_Preferred
Molecular Formula C41H70O9 • Ca, Purity ≥98%, Formula Weight 747.1; A selective calcium ionophore that mobilizes intracellular calcium stores. It is used as a research tool to raise the intracellular level of calcium, to study calcium transport across biological membranes, and to stimulate the intracellular production of cytokines.
Catalog No. NC1486026