Learn More
Research Products International Corp Ionomycin Free Acid, 1 Milligrams
SDP
Supplier:  Research Products International Corp I520200.001
Encompass_Preferred
Ionomycin free acid is frequently employed in cell biology experiments to study cellular processes that are dependent on calcium signaling. By transporting calcium ions into cells, ionomycin can trigger various cellular responses, including the activation of enzymes and signaling pathways. 
Researchers use ionomycin in a variety of applications, such as studying cell signaling, apoptosis, and calcium-dependent processes. It is particularly useful in experiments where precise control over intracellular calcium levels is necessary.
Catalog No. 50-488-721