Learn More
AdipoGen Ionomycin (free acid),Chemical. CAS: 56092-81-0.
SDP
Supplier:  AdipoGen AGCN20416M005
Encompass_Preferred
Ionomycin (free acid), AdipoGen Life Sciences. Chemical. CAS: 56092-81-0. Formula: C41H72O9. MW: 709. Isolated from Streptomyces conglobatus. Antibiotic. Potent and highly selective Ca2+ ionophore. Commonly used to modify intracellular calcium levels to study calcium transport across biological membranes and to calibrate fluorescent Ca2+ indicators. Ionomycin also transports Pb2+ and some other divalent cations, as well as several lanthanide series trivalent cations at efficiencies that are greater than or equal to those for Ca2+. Apoptosis inducer. Induces apoptotic neuronal degeneration in embryonic cortical neurons and cell cycle arrest at G1 phase and induces central demyelination. Used to stimulate the intracellular production of the cytokines, interferon, perforin, IL-2 and IL-4 usually in conjunction with PMA. ADAM10 agonist. Potent inducer of shedding CXCL16. TREK-1 channels inhibitor.
Catalog No. 50-596-348