Learn More
Medchemexpress LLC Ionotropic Glutamate Receptor 2 Antibody (YA1143) | 100 UL

Supplier: Medchemexpress LLC HYP81398100UL
The Ionotropic Glutamate Receptor 2 Antibody (YA1143) is a rabbit-derived, non-conjugated IgG monoclonal antibody targeting Ionotropic Glutamate Receptor 2 in human. Glutamate receptors are the primary excitatory neurotransmitter receptors in the mammalian brain, involved in various neurophysiologic processes. This product is intended for research use only.
- Host: Rabbit
- Reactivity: Human
- Conjugation: Non-conjugated
- Formulation: Supplied in PBS with 50% glycerol, 0.5% BSA, and 0.02% sodium azide (pH 7.139)
- Purity: Affinity purified
- Storage: Stored at -20°C for 1 year. Avoid repeated freeze/thaw cycles
- Appearance: Liquid
- Application: IHC-P
- Dilution ratio: 1:100-1:200
- Molecular weight: 99 kDa
- Concentration: 0.8 mg/mL
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.