Learn More
Medchemexpress LLC Iopamidol | 60166-93-0 | 99.9% | 500 MG
SDP
Supplier:  Medchemexpress LLC HYB0684500MG
Encompass_Preferred
Iopamidol is a nonionic, X-Ray iodinated contrast agent (CA) used for various diagnostic applications. It contains amide and hydroxyl functionalities that can be used for generating chemical exchange saturation transfer (CEST) contrast. Iopamidol can also serve as a responsive MRI-chemical exchange saturation transfer contrast agent for pH mapping of kidneys.
  • Nonionic, X-ray iodinated contrast agent
  • Used for various diagnostic applications
  • Contains amide and hydroxyl functionalities for chemical exchange saturation transfer (CEST) contrast
  • Can be used for pH mapping of kidneys
  • High purity (99.9%)
  • White to off-white solid appearance
Catalog No. 50-163-9571