Learn More
Medchemexpress LLC Iopamidol | 60166-93-0 | 99.9% | 1 ML
SDP
Supplier:  Medchemexpress LLC HYB068410MM/1ML
Encompass_Preferred
Iopamidol is a nonionic, X-Ray iodinated contrast agent (CA) used for a wide variety of diagnostic applications. It contains amide and hydroxyl functionalities that can be exploited for the generation of chemical exchange saturation transfer (CEST) contrast.
  • Nonionic, X-ray iodinated contrast agent
  • Used for various diagnostic applications
  • Contains amide and hydroxyl functionalities for CEST contrast generation
  • Can be used for pH mapping of kidneys as a responsive MRI-CEST contrast agent
Catalog No. 50-004-09182