Learn More
Medchemexpress LLC Iopamidol | 60166-93-0 | 99.9% | 50 MG
SDP
Supplier:  Medchemexpress LLC HYB068450MG
Encompass_Preferred
Iopamidol is a nonionic, X-Ray iodinated contrast agent (CA) suitable for various diagnostic applications. It contains amide and hydroxyl functionalities that can be utilized for generating chemical exchange saturation transfer (CEST) contrast. This product is for research use only.
  • Nonionic, X-Ray iodinated contrast agent
  • Suitable for a wide variety of diagnostic applications
  • Amide and hydroxyl functionalities can be exploited for generating chemical exchange saturation transfer (CEST) contrast
  • Can be used as a responsive MRI-chemical exchange saturation transfer contrast agent for pH mapping of kidneys in vivo
Catalog No. 50-004-08438