Learn More
Medchemexpress LLC Iophenoxic acid | 96-84-4 | MFCD00133419 | 98.2% | 571.92 g/mol | C11H11I3O3 | 5 G
SDP
Supplier:  Medchemexpress LLC HYW2335055G
Encompass_Preferred
Iophenoxic acid is an iodinated biochemical reagent used as a systemic blood marker to assess bait acceptance in small mustelids, such as stoats and weasels. It is provided for research and analytical applications where reliable marker detection is required.
  • Used as a systemic blood marker for bait acceptance studies.
  • Iodinated small molecule suitable for analytical assays.
  • High purity for reliable experimental results.
  • Molecular weight 571.92 g/mol; formula C11H11I3O3.
  • Available in laboratory-scale pack sizes for research use.
Catalog No. 50-002-82261