Learn More
BIOLIFE SOLUTIONS INC ThawSTAR IOP Qualification Packet
SDP
Supplier:  BIOLIFE SOLUTIONS INC AST614
Encompass
The IOPQ Packet includes more than 100 pages of documentation and testing tool accessories to facilitate functional (“Black Box”) testing of the instrument. Integration of ThawSTAR CFT2 Transporter during the qualification process is optional, and depends on your “intended use”. Protocol templates are customizable to meet organizational needs. Appendices, data sheets, and other support documents are all included in one complete binder.
Catalog No. NC1644386