Learn More
Medchemexpress LLC Iota-carrageenan | 9062-07-1 | MFCD00151512 | 98.0% | No defined molecular weight (polymeric) | No single defined chemical formula (polymeric) | 5 G
SDP
Supplier:  Medchemexpress LLC HYW1455235G
Encompass_Preferred
Iota-carrageenan is an iota-type sulfated polysaccharide supplied as a powder for laboratory research. It is used as a biochemical assay reagent and as a gelling or proinflammatory agent in life-science studies. The material is intended for research use only and is provided with supporting documentation for safe handling and quality control.
  • Is an iota-type sulfated polysaccharide extracted from red seaweed.
  • Functions as a biochemical assay reagent and proinflammatory agent in research.
  • Forms thermoreversible gels and can modify viscosity in formulations.
  • Supplied as a powder with listed purity of 98.0%.
  • Available in small pack sizes suitable for laboratory use (e.g., 5 G).
  • Storage: powder stable at -20°C (3 years) or 4°C (2 years); in solvent stability is shorter.
  • COA, SDS, and datasheet are available for handling and quality information.
Catalog No. 50-002-83717