Learn More
Medchemexpress LLC Ioversol | 87771-40-2 | 98.0% | 50 MG
SDP
Supplier:  Medchemexpress LLC HYB141050MG
Encompass_Preferred
Ioversol is a nonionic iodinated contrast medium used during CT scans or x-rays in animal experiments. It does not damage the blood-brain barrier in animals.
  • Nonionic iodinated contrast medium.
  • Used in CT scans or x-rays in animal experiments.
  • Does not damage the blood-brain barrier (BBB) in animals.
  • Reduced tendency to stimulate endothelin release compared to iothalamate.
  • Produces less renal vasoconstriction than iothalamate in control and USIC rats.
  • Helps avoid the development of radiocontrast nephropathy.
Catalog No. 50-004-12577