Learn More
Medchemexpress LLC Ioversol | 87771-40-2 | MFCD00867959 | 98.0% | 807.11 g/mol | C18H24I3N3O9 | 10 MG

Supplier: Medchemexpress LLC HYB141010MG
Ioversol is a non-ionic, low-osmolar iodinated contrast agent used to enhance CT and x-ray imaging in preclinical (animal) studies. It is water-soluble, supplied for research use only, and characterized by high iodine content for radiographic contrast.
- Non-ionic iodinated contrast agent for CT and x-ray imaging.
- Purity 98.0%.
- Molecular weight 807.11 g/mol.
- Chemical formula C18H24I3N3O9.
- CAS number 87771-40-2.
- Available as a 10 mg research package.
- Does not damage the blood-brain barrier in animal studies.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.