Learn More
Medchemexpress LLC IOWH-032 | 1191252-49-9 | 99.6% | 545.18 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY183375MG
Encompass_Preferred
IOWH-032 is a synthetic anti-secretory molecule and a potent CFTR inhibitor with an IC50 value of 8 μM. It also functions as an anti-diarrheal agent.
  • Potent CFTR inhibitor with an IC50 value of 8 μM.
  • Acts as an anti-diarrheal agent.
  • Increases ACE-2 expression in SARS-CoV-2 infected CFBE41o- WT cells.
Catalog No. 50-003-87581