Learn More
Medchemexpress LLC Ioxilan | 107793-72-6 | MFCD00867280 | 98.0% | 791.11 g/mol | C18H24I3N3O8 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1095135MG
Encompass_Preferred
Ioxilan is a low-osmolar, nonionic, iodinated contrast agent provided as a research-grade reference compound for analytical, formulation, and preclinical studies. It is supplied as a purified, water-soluble solid suitable for method development and small-scale laboratory use.
  • Low-osmolar, nonionic iodinated contrast agent.
  • High purity suitable for analytical and preclinical research.
  • Available in small milligram packages for laboratory use.
  • Water-soluble, compatible with common formulation solvents.
  • Stable when stored under recommended conditions.
Catalog No. 50-002-64718