Learn More
Medchemexpress LLC Ioxitalamic acid | 28179-44-4 | MFCD00867942 | 97.0% | 643.94 g/mol | C12H11I3N2O5 | 5MG
SDP
Supplier:  Medchemexpress LLC HY1256955MG
Encompass_Preferred
Ioxitalamic acid is a tri-iodinated, water-soluble iodinated contrast agent used in X-ray and CT imaging research. It is described as nephrophilic and highly permeable and is supplied as a solid for biochemical assay and research use. Key identifiers include CAS 28179-44-4, molecular formula C12H11I3N2O5, molecular weight 643.94 g/mol, and typical purity of 97.0%.
  • Water-soluble tri-iodinated contrast agent.
  • High iodine content for strong radiopacity.
  • Nephrophilic and highly permeable.
  • Supplied as a solid suitable for research assays.
  • Available in small research quantities such as 5 mg.
  • Typical purity reported as 97.0%.
Catalog No. 50-002-39361