Learn More
Medchemexpress LLC IP-10/CRG-2/CXCL10 5ug
SDP
Supplier:  Medchemexpress LLC HYP72275UG
Encompass_Preferred
CXCL10 also known as interferon -induced protein 10 kDa (IP-10) is a cytokine belonging to the CXC chemokine family CXCL10 exerts its biological effects by binding to CXCR3 CXCL10 is a pleiotropic molecule capable of exerting potent biological functions including promoting the chemotactic activity of CXCR3 cells inducing apoptosis regulating cell growth and proliferation as well as angiogenesis in infectious and inflammatory diseases and cancer[1] IP-10/CRG-2/CXCL10 Protein Mouse consists of 77 amino acids (I22-P98) and is expressed in E coli
Catalog No. 50-001-81945