Learn More
Cayman Chemical IP Wash BuFR DeTRgent10X 50mL
SDP
Supplier:  Cayman Chemical 60197350 ML
Encompass_Preferred
IP Wash Buffer Detergent (10X) has been tested and formulated to work exclusively with Cayman’s Protein A/G Coated Plate Immunoprecipitation Kit (Item No. 601970)
Catalog No. 50-289-6368