Learn More
Abcam IP10 (CXCL10) Antibody Pair - BSA and Azide free, 10 x 96 Tests
Supplier:  Abcam AB25384610X96TESTS
Encompass_Preferred
IP10 (CXCL10) Antibody Pair - BSA and Azide free is a kit containing recombinant capture and detector antibodies in a carrier-free formulation for the measurement of IP10 (CXCL10).
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-265-7017