Learn More
Selleck Chemical LLC IPA-3 5mg 42521-82-4
SDP
Supplier:  Selleck Chemical LLC S70935MG
Encompass_Preferred
IPA-3 is a selective non-ATP competitive Pak1 inhibitor with IC50 of 2.5 ?M in a cell-free assay, no inhibition to group II PAKs (PAKs 4-6). *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-4392