Learn More
Medchemexpress LLC HY-15663 5mg Medchemexpress, IPA-3 CAS:42521-82-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY156635MG
Encompass_Preferred
Medchemexpress, HY-15663 5mg IPA-3 CAS:42521-82-4 IPA-3 is a selective non-ATP competitive PAK1 inhibitor with IC50 of 2.5 μM, and shows no inhibition to group II PAKs (PAKs 4-6). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0664