Learn More
Medchemexpress LLC Ipag | 193527-91-2 | 99.8% | 395.28 g/mol | C17H22IN3 | 10 MG

Supplier: Medchemexpress LLC HY10098510MG
IPAG is a research-grade sigma-1 receptor antagonist reported to have a pKi of 4.3 and shown to induce apoptosis in cellular models. Supplied as a solid for laboratory use, the compound is characterized by a defined molecular formula and high purity suitable for biochemical and pharmacological studies.
- Potent sigma-1 receptor antagonist (pKi = 4.3).
- Shown to induce apoptosis in cell models.
- High purity suitable for research applications (≈99.8%).
- Stable as a solid when stored at 4°C, protected from light.
- Available in milligram-scale pack sizes for laboratory use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.