Learn More
Medchemexpress LLC Ipatasertib | 1001264-89-6 | C24H32ClN5O2 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY1518610MM/1ML
Encompass_Preferred
Ipatasertib (GDC-0068) is an orally active, highly selective, and ATP-competitive pan-Akt inhibitor with IC50 values of 5, 18, and 8 nM for Akt1, Akt2, and Akt3, respectively. It synchronously activates FoxO3a and NF-κB by inhibiting Akt, leading to p53-independent activation of PUMA. Ipatasertib also induces apoptosis in cancer cells and inhibits tumor growth in xenograft mouse models.
  • Orally active
  • Highly selective and ATP-competitive pan-Akt inhibitor
  • Induces apoptosis in cancer cells
  • Inhibits tumor growth in xenograft mouse models
  • Purity: 99.79%
Catalog No. 50-003-74854