Learn More
Medchemexpress LLC Ipatasertib | 1001264-89-6 | C24H32ClN5O2 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY15186
Encompass_Preferred
Ipatasertib (GDC-0068) is an orally active, highly selective, and ATP-competitive pan-Akt inhibitor with IC50 values of 5, 18, and 8 nM for Akt1, Akt2, and Akt3, respectively. It synchronously activates FoxO3a and NF-κB through Akt inhibition, leading to p53-independent activation of PUMA. Ipatasertib also induces apoptosis in cancer cells and inhibits tumor growth in xenograft mouse models.
  • Purity 99.79%
  • Targets Akt1 (5 nM), Akt2 (18 nM), Akt3 (8 nM), and PKA (3100 nM)
  • Demonstrates antiproliferative activity against various human cell lines
  • Suppresses colon cancer cell proliferation by activating PUMA in vitro
  • Exhibits PUMA-dependent antitumor activity in xenograft mouse models
Catalog No. 50-003-75994