Learn More
Medchemexpress LLC HY-15186A 5mg Medchemexpress, Ipatasertib dihydrochloride CAS:1396257-94-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY15186A5MG
Encompass_Preferred
Medchemexpress, HY-15186A 5mg Ipatasertib dihydrochloride CAS:1396257-94-5 Ipatasertib dihydrochloride (GDC-0068 dihydrochloride) is a highly selective and ATP-competitive pan-Akt inhibitor with IC50s of 5, 18 and 8 nM for Akt1, Akt2 and Akt3, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0589