Learn More
Medchemexpress LLC Ipatasertib-NH2 | 1001382-14-4 | 99.6% | 10 MM 1 ML
SDP
Supplier:  Medchemexpress LLC HY13098810MM/1ML
Encompass_Preferred
Ipatasertib-NH2 is a ligand targeting the AKT protein, utilized in the PROTAC (Proteolysis Targeting Chimera) INY-03-041. This compound plays a role in the selective degradation of target proteins by exploiting the ubiquitin-proteasome system, and is composed with a ten-hydrocarbon linker and a CRBN ligand Lenalidomide. This product is for research use only.
  • Functions as a ligand for target protein AKT
  • Utilized in proteolysis targeting chimera (PROTAC) applications
  • Designed for selective degradation of target proteins
  • Contains a ten-hydrocarbon linker and a CRBN ligand
Catalog No. 50-003-40013