Learn More
Selleck Chemical LLC Eganelisib (IPI-549) 5mg 1693758-51-8
SDP
Supplier:  Selleck Chemical LLC S83305MG
Encompass_Preferred
Eganelisib (IPI-549) is a potent inhibitor of PI3K-? with >100-fold selectivity over other lipid and protein kinases. The biochemical IC50 for PI3K-? is 16 nM. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8007