Learn More
Selleck Chemical LLC Ipilimumab (anti-CTLA-4) 1mg 477202-00-9 MDX-010, BMS-734016, Yervoy
SDP
Supplier:  Selleck Chemical LLC A20011MG
Encompass_Preferred
Ipilimumab (anti-CTLA-4) is an immunomodulatory monoclonal antibody directed against the cell surface antigen CTLA-4 and also a type of immune checkpoint inhibitor. MW : 148 kD. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-0843