Learn More
Cayman Chemical IpraglIflozIn-13C6 1mg
SDP
Supplier:  Cayman Chemical 287051 MG
Encompass_Preferred
An internal standard for the quantification of ipragliflozin by GC- or LC-MS
Catalog No. 50-293-8245