Learn More
Medchemexpress LLC Ipragliflozin | 761423-87-4 | 99.7% | 404.45 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY14894100MG
Encompass_Preferred
Ipragliflozin is an orally active and selective SGLT2 inhibitor that functions as an antidiabetic agent. It has been shown to exhibit potent inhibitory activity against human, rat, and mouse SGLT2, and is intended for research purposes only.
  • Selective SGLT2 inhibitor
  • Demonstrates antihyperglycemic effects
  • Suppresses the growth of MCF-7 human breast cancer cell lines in vitro
  • Inhibits DNA synthesis in MCF-7 cells in vitro
  • Reduces HbA1c and blood glucose levels in vivo
  • Increases pancreatic insulin content in vivo
  • Increases urinary glucose excretion in vivo
Catalog No. 50-003-60905