Learn More
Medchemexpress LLC Ipragliflozin (L-Proline) | 951382-34-6 | 99.9% | 519.58 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY14894A50MG
Encompass_Preferred
Ipragliflozin (L-Proline) is a highly potent and selective SGLT2 inhibitor with an IC50 of 2.8 nM. It shows little to no potency for SGLT1/3/4/5/6.
  • Potently and selectively inhibits human, rat, and mouse SGLT2 at nanomolar ranges.
  • Exhibits stability against intestinal glucosidases.
  • Demonstrates good pharmacokinetic properties after oral dosing.
  • Dose-dependently increases urinary glucose excretion, with effects lasting over 12 hours in normal mice at doses of 0.3 mg/kg or higher.
  • Single administration dose-dependently increases urinary glucose excretion, reduces blood glucose and plasma insulin levels, and improves glucose intolerance.
Catalog No. 50-003-60907