Learn More
Medchemexpress LLC IPrAuCl | 852445-83-1 | 622.02 | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHYW010710
Encompass_Preferred
IPrAuCl is a gold compound known for its inhibitory activity against sarcoplasmic reticulum Ca²⁺-ATPase, with an IC₅₀ of 16.3 μM. This product is supplied as a white to off-white solid, tested to ensure consistency with its structure and compliance with elemental analysis specifications for laboratory research.
  • Inhibits sarcoplasmic reticulum Ca²⁺-ATPase (IC₅₀=16.3 μM)
  • Provided as a white to off-white solid
  • Consistent with structure by 1H NMR spectrum
  • Complies with elemental analysis specifications
Catalog No. 50-004-53314