Learn More
Medchemexpress LLC Ipriflavone | 35212-22-7 | MFCD00221719 | 100.0% | 280.32 g/mol | C18H16O3 | 10 MG

Supplier: Medchemexpress LLC HYN0094R10MG
Ipriflavone analytical standard provided for research and analytical applications as a reference material for assay development and quality control. The compound is a synthetic isoflavone derivative supplied as a high-purity solid suitable for analytical workflows.
- High purity: 99.95%.
- Chemical formula: C18H16O3.
- Molecular weight: 280.32 g/mol.
- Appearance: white to off-white solid.
- Intended use: reference standard for research and analytical purposes.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.