Learn More
Cayman Chemical IpronIdazole-d3 1mg
SDP
Supplier:  Cayman Chemical 371941 MG
Encompass_Preferred
An internal standard for the quantification of ipronidazole by GC- or LC-MS
Catalog No. 50-291-5135