Learn More
Cellular Engineering Technologies iPSC Passaging Solution (100mL)
Supplier:  Cellular Engineering Technologies MR1002
Encompass_Preferred
This xeno-free and feeder-free media is uniquely formulated to grow and expand induced pluripotent stem cells. This formulation was developed and refined over decades of bench work using CET's proprietary cell lines. This media has been shown to support iPSC growth for up to 20 passages without significant colony variation. Media is shipped with gel packs.
Catalog No. NC2753059