Learn More
Cellular Engineering Technologies iPS Reprogramming Media Kit (iPSC Base Media 250mL and iPSC Reprogramming Supplement 500µL)
Supplier:  Cellular Engineering Technologies MR1003K
Encompass_Preferred
Specifically designed to increase the efficiency of all iPS cell reprogramming methods that are currently utilized. This kit requires a media change every 48 hours, saving researchers hands-on time and media consumption. Target cells that are mitotically active and in a low passage state are highly recommended. Base media is shipped with gel packs. A separate growth supplement is shipped on dry ice.Components:iPS Base Media 250mLiPS Reprogramming Supplement 500µL
Catalog No. NC2753060
Only null left
Add to Cart
Request a Quote