Learn More
Medchemexpress LLC Iptacopan | 1644670-37-0 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY127105
Encompass_Preferred
Iptacopan (LNP023) is an effective, orally-active, highly selective factor B inhibitor with an IC50 value of 10 nM and a KD of 7.9 nM. It exerts a proximal effect in the complement cascade reaction, preventing the destruction (hemolysis) of red blood cells in Paroxysmal Nocturnal Hemoglobinuria (PNH) and the damage of renal cells in Immunoglobulin A Nephropathy (IgAN) and C3 glomerulopathy (C3G). Iptacopan can be used for the study of complement-mediated diseases, particularly PNH, primary IgAN, and C3G.
  • Effective and orally-active
  • Highly selective factor B inhibitor
  • Has an IC50 value of 10 nM and a KD of 7.9 nM
  • Exerts a proximal effect in the complement cascade
Catalog No. 50-003-41727