Learn More
Medchemexpress LLC Iptacopan (hydrochloride) | 1646321-63-2 | 99.83% | 458.98 | 5 MG
SDP
Supplier:  Medchemexpress LLC 5MGHY127105A
Encompass_Preferred
Iptacopan hydrochloride is an effective, orally-active, and highly selective factor B inhibitor. It acts proximally in the complement cascade, preventing red blood cell destruction in paroxysmal nocturnal hemoglobinuria (PNH) and renal cell damage in immunoglobulin A nephropathy (IgAN) and complement 3 glomerulopathy (C3G).
  • Effective, orally-active, and highly selective factor B inhibitor
  • IC50 value of 10 nM and KD of 7.9 nM for factor B
  • Prevents red blood cell destruction in PNH
  • Prevents renal cell damage in IgAN and C3G
  • Inhibits alternative complement pathway (AP)-induced membrane attack complex (MAC) formation
  • Exhibits excellent selectivity over other proteases
  • Used for studying complement-mediated diseases
Catalog No. 50-003-64374