Learn More
TARGETMOL CHEMICALS INC IPTACOPAN HYDROCHLORIDE 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T91895MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Iptacopan hydrochloride (LNP023 hydrochloride) is an orally bioavailable highly potent and highly selective factor B inhibitor with an IC50 of 10 nM. Iptacopan hydrochloride shows direct reversible and high-affinity binding to human factor B with a KD of 7.9 nM. purity: 99%
Catalog No. 50-257-9013