Learn More
Medchemexpress LLC Iptakalim hydrochloride | 642407-63-4 | 98.0% | 179.73 g/mol | C9H22ClN | 10 MG

Supplier: Medchemexpress LLC HY10806910MG
Iptakalim hydrochloride is a lipophilic para-amino small molecule used in research as an ATP-sensitive potassium (KATP) channel opener and α4β2-containing nicotinic acetylcholine receptor modulator. It is supplied as the hydrochloride salt for laboratory pharmacology and formulation studies.
- Acts as an ATP-sensitive potassium (KATP) channel opener and α4β2-containing nicotinic receptor modulator.
- CAS number 642407-63-4.
- Molecular formula C9H22ClN; molecular weight 179.73 g/mol.
- Purity 98.0%.
- Available in milligram pack sizes for research, including 10 mg.
- Solubility: water 25 mg/mL (requires warming and sonication); PBS formulations achievable.
- Storage: solid sealed at 4°C; in solution store at -80°C for long-term stability.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.