Learn More
Research Products International Corp Isopropyl-B-D-thio-galactopyranoside [IPTG], 100 Grams
SDP
Supplier:  Research Products International Corp I56000100.0
Encompass_Preferred
IPTG is combined with X-GAL to detect lac+ colonies or cells in a colorimetric assay. Distinguishes recombinants (white) from non- recombinants (blue) in cloning strategies using vectors such as Lambda-11, M13mp 18 and 19, pUC18 and 19, pUR222 containing the lacZ gene.
Dioxane-Free
Water Content: <0.49%
Catalog No. 50-213-378