Learn More
Medchemexpress LLC HY-15921 10g , IPTG CAS:367-93-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1592110G
Encompass_Preferred
HY-15921 10g IPTG CAS: 367-93-1 IPTG is a molecular mimic of allolactose, a lactose metabolite that triggers transcription of the lac operon, and it is therefore used to induce protein expression where the gene is under the control of the lac operator. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0960