Learn More
Biotang Inc IPTG 25 GM, P-806, CAS # 367-93-1. Purity > 99%, soluble in ethanol and water. Shipped at room temperature. Store at 2-8 degrees Celsius. Dioxane free. Mol. wt. 238.3.
Supplier:  Biotang Inc BTBB903
Encompass_Preferred
IPTG 25 GM, P-806, CAS # 367-93-1. Purity > 99%, soluble in ethanol and water. Shipped at room temperature. Store at 2-8 degrees Celsius. Dioxane free. Mol. wt. 238.3.
Catalog No. 50-152-357