Learn More
Research Products International Corp IPTG [Isopropyl-β-D-thiogalactopyranoside], Dioxane Free, 50 Grams
SDP
Supplier:  Research Products International Corp I5600050.0
Encompass_Preferred
IPTG is combined with X-GAL to detect lac+ colonies or cells in a colorimetric assay. Distinguishes recombinants (white) from non- recombinants (blue) in cloning strategies using vectors such as Lambda-11, M13mp 18 and 19, pUC18 and 19, pUR222 containing the lacZ gene.
Dioxane-Free
Water Content: <0.49%
Catalog No. 50-136-8310