Learn More
Teknova 1M IPTG. 50mL, Sterile.
SDP
Supplier:  Teknova I3430
Encompass
IPTG (isopropyl-beta-D-thiogalactopyranoside) is a galactose analog that can be used to induce expression of cloned genes under control of the lac operon. It is often used in conjunction with X-Gal or Bluo-Gal in blue-white selection of recombinant bacterial colonies.
Catalog No. 50-841-757