Learn More
Medchemexpress LLC IQ-1S free acid | 143534-71-2 | >98.0% | 385.39 g/mol | C18H15N3O5S | 1 ML

Supplier: Medchemexpress LLC HY10023310MM 1ML
IQ-1S free acid is a research-grade small-molecule inhibitor reported to suppress NF-κB and AP-1 activity and to bind JNK isoforms. It is provided as a powder and as a ready-to-use 10 mM solution in DMSO for laboratory research.
- Research-grade NF-κB and AP-1 inhibitor with documented JNK binding.
- Available as powder and 10 mM solution in DMSO for convenient use.
- High purity >98% suitable for biochemical and cell-based assays.
- Molecular formula C18H15N3O5S; molecular weight 385.39 g/mol.
- Powder form recommended to be stored desiccated at -20°C; follow solvent storage guidelines for solutions.
- Supplied in multiple pack sizes to support a range of experimental scales.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.